missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylsulfoxide, 99.9% min., OmniSolv, MilliporeSigma™
Supplier: Technidata MX14561
Specifications
| MethylSulfoxide | |
| 67-68-5 | |
| 1.10g/cm3 at 20°C | |
| 4 L | |
| 87°C | |
| Glass Bottle | |
| 99.9% min. | |
| (CH3)2SO | |
| 0.053hPa (20°C) | |
| IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
| methanesulfinylmethane | |
| 679 | |
| 78.1 |
| 18.4°C | |
| Colorless | |
| 189°C | |
| 1.8 to 63.0%(V) | |
| 214.85°C (418.7°F) | |
| Liquid | |
| C2H6OS | |
| MFCD00002089 | |
| dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
| CS(C)=O | |
| 78.13 | |
| CHEBI:28262 |
Chemical Identifiers
| 67-68-5 | |
| 78.13 | |
| IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
| 679 | |
| methanesulfinylmethane |
| C2H6OS | |
| MFCD00002089 | |
| dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
| CHEBI:28262 | |
| CS(C)=O |
Safety and Handling
Recommended Storage : Store at +15°C to +25°C