missing translation for 'onlineSavingsMsg'
Learn More

Methylsulfoxide, 99.8% min., DriSolv™, MilliporeSigma™

Catalog No. MMX14576
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MMX14576 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MMX14576 Supplier Technidata Supplier No. MX14576

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methanesulfinylmethane
SMILES CS(C)=O

Specifications

Quantity 1 L
Physical Form Liquid
Assay Percent Range 99.8%, min.
Linear Formula (CH3)2SO
Formula Weight 78.14
Chemical Name or Material Methylsulfoxide, DriSolv™
Recommended Storage Room Temperature