missing translation for 'onlineSavingsMsg'
Learn More

Methylprednisolone Acetate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014505
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014505 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014505 Supplier Merck Emd Millipore Supplier No. PHR1718500MG

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Methylprednisolone is a synthetic corticosteroid drug which is employed for the management of inflammatory diseases. It is used to relieve the inflammations caused by psoriasis, eczema, arthritis, asthma and rhinitis. It is also widely employed for the treatment of acute lung injury caused by acid aspiration.

Specifications

CAS 53-36-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C24H32O6
Synonym 11 beta,17,21-Trihydroxy-6 alpha-methylpregna-1,4-diene-3,20-dione 21-acetate
Type Certified Reference Material
Quantity 500 mg
Formula Weight 416.51
Grade Certified Reference Material