missing translation for 'onlineSavingsMsg'
Learn More

Methylparaben, Spectrum™ Chemical

Catalog No. 18611808
Change view
Click to view available options
Quantity:
100 g
500 g
5 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611808 500 g Poly Bottle
18611805 100 g Poly Bottle
18611806 5 kg Poly Bottle
18611807 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18611808 Supplier Spectrum Chemical Mfg Cor Supplier No. H2265500GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 99-76-3

Specifications

CAS Max % 1
Boiling Point 270 to 280°C
Packaging Poly Bottle
Quantity 500 g
Grade Reagent
Physical Form Crystalline Powder or Lumps

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.