missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylparaben Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Methylparaben is an antifungal and antibacterial agent, widely used as a preservative in cosmetics, pharmaceuticals, food and beverages.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 99-76-3 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available. |
| Physical Form | Neat |
| Linear Formula | HOC6H4 CO2 CH3 |
| MDL Number | MFCD00002352 |
| Melting Point | 125°C to 128°C |
| Synonym | Methyl 4-hydroxybenzoate; p-Hydroxybenzoic acid methyl ester; Methyl paraben; NIPAGIN |
| Type | Certified Reference Material |
| Quantity | 1 g |
| Formula Weight | 152.15 |
| Show More |