missing translation for 'onlineSavingsMsg'
Learn More

Methylparaben Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111016024
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016024 1 g
111016025 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111016024 Supplier Merck Emd Millipore Supplier No. PHR10121G
Methylparaben is an antifungal and antibacterial agent, widely used as a preservative in cosmetics, pharmaceuticals, food and beverages.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications

CAS 99-76-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula HOC6H4 CO2 CH3
MDL Number MFCD00002352
Melting Point 125°C to 128°C
Synonym Methyl 4-hydroxybenzoate; p-Hydroxybenzoic acid methyl ester; Methyl paraben; NIPAGIN
Type Certified Reference Material
Quantity 1 g
Formula Weight 152.15
Grade Certified Reference Material
Show More Show Less