missing translation for 'onlineSavingsMsg'
Learn More

Methylparaben, NF, 98-102%, Spectrum™ Chemical

Catalog No. 18604534
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
12 kg
2.5 kg
25 g
50 kg
500 g
Packaging:
Poly Pail
Catalog No. Quantity Packaging
18604534 12 kg Poly Pail
18604535 25 g Poly Pail
18604536 100 g Poly Pail
18604532 500 g Poly Pail
18604533 2.5 kg Poly Pail
18604537 50 kg Poly Pail
6 options

Catalog No. 18604534

Supplier: Spectrum Chemical Mfg Cor ME16312KG

Only null left

ME163, 99-76-3, C8H8O3

Spectrum™ Chemical Methylparaben, NF is an excipient used as a preservative. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 99-76-3
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
InChI Key LXCFILQKKLGQFO-UHFFFAOYSA-N
IUPAC Name methyl 4-hydroxybenzoate
SMILES COC(=O)C1=CC=C(O)C=C1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Packaging Poly Pail
Quantity 12 kg
Percent Purity 98 to 102%
Grade NF
Physical Form Crystalline Powder or Lumps