Learn More
Methylmagnesium bromide, 1M solution in CPME, AcroSeal™, Thermo Scientific™
$144.14 - $948.25
Chemical Identifiers
| CAS | 75-16-1 , 5614-37-9 |
|---|---|
| Molecular Formula | CH3BrMg |
| Molecular Weight (g/mol) | 119.24 |
| MDL Number | MFCD00000041 |
| InChI Key | NXPHGHWWQRMDIA-UHFFFAOYSA-M |
| Synonym | methylmagnesium bromide, methyl magnesium bromide, magnesium, bromomethyl, bromo methyl magnesium, methylmagnesium bromide solution, 3.0 m in diethyl ether, unii-22cw9773df, memgbr, methymagnesiumbromide, ch3mgbr, methylmagnesiumbromide, grignard reagent |
| PubChem CID | 6349 |
| IUPAC Name | magnesium;carbanide;bromide |
| SMILES | [CH3-].[Mg++].[Br-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC446001000
|
N/A | 100mL |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC446008000
|
N/A | 800mL |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| CH3BrMg | |
| MFCD00000041 | |
| methylmagnesium bromide, methyl magnesium bromide, magnesium, bromomethyl, bromo methyl magnesium, methylmagnesium bromide solution, 3.0 m in diethyl ether, unii-22cw9773df, memgbr, methymagnesiumbromide, ch3mgbr, methylmagnesiumbromide, grignard reagent | |
| magnesium;carbanide;bromide |
Specifications
| CH3BrMg | |
| MFCD00000041 | |
| methylmagnesium bromide, methyl magnesium bromide, magnesium, bromomethyl, bromo methyl magnesium, methylmagnesium bromide solution, 3.0 m in diethyl ether, unii-22cw9773df, memgbr, methymagnesiumbromide, ch3mgbr, methylmagnesiumbromide, grignard reagent | |
| NXPHGHWWQRMDIA-UHFFFAOYSA-M | |
| magnesium;carbanide;bromide | |
| 6349 | |
| Methylmagnesium bromide |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapour.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful to aquatic life with
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomi
GHS Signal Word: Danger
EINECSNumber : 200-844-1
RUO – Research Use Only