missing translation for 'onlineSavingsMsg'
Learn More

Methylmagnesium Bromide (12% in Tetrahydrofuran, ca. 1mol/L), TCI America™

Catalog No. M0362250G
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0362250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M0362250G Supplier TCI America Supplier No. M0362250G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75-16-1
Molecular Formula CH3BrMg
Molecular Weight (g/mol) 119.24
MDL Number MFCD00000041
InChI Key AVFUHBJCUUTGCD-UHFFFAOYSA-M
Synonym methylmagnesium bromide, methyl magnesium bromide, magnesium, bromomethyl, bromo methyl magnesium, methylmagnesium bromide solution, 3.0 m in diethyl ether, unii-22cw9773df, memgbr, methymagnesiumbromide, ch3mgbr, methylmagnesiumbromide
PubChem CID 6349
IUPAC Name bromo(methyl)magnesium
SMILES C[Mg]Br

Specifications

UN Number 3399
Formula Weight 119.24
Quantity 250 g
Chemical Name or Material Methylmagnesium Bromide (12% in Tetrahydrofuran, ca. 1mol/L)
EINECSNumber (2)-3468
TSCA Yes