missing translation for 'onlineSavingsMsg'
Learn More

Methylglyoxime 98.0+%, TCI America™

Catalog No. M050725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M050725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M050725G Supplier TCI America Supplier No. M050725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1804-15-5
Molecular Formula C3H6N2O2
Molecular Weight (g/mol) 102.093
MDL Number MFCD00059608
InChI Key YYWZDZMCXPSGDF-NSCUHMNNSA-N
PubChem CID 5355463
IUPAC Name N-[(E)-1-nitrosoprop-1-en-2-yl]hydroxylamine
SMILES CC(=CN=O)NO

Specifications

Melting Point 157°C
Color White
Quantity 25 g
Formula Weight 102.09
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Methylglyoxime