accessibility menu, dialog, popup

UserName

CAS RN 1804-15-5

IUPAC Name: N-[(1E)-1-nitrosoprop-1-en-2-yl]hydroxylamine
Synonyms: N-[(1E)-1-nitrosoprop-1-en-2-yl]hydroxylamine
Molecular Weight (g/mol): 102.09
Molecular Formula: C3H6N2O2
InChi Key: YYWZDZMCXPSGDF-NSCUHMNNSA-N
SMILES: C\C(NO)=C/N=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results