missing translation for 'onlineSavingsMsg'
Learn More

Methylenediphenyl 4,4'-Diisocyanate 97.0+%, TCI America™

Catalog No. p-7126488
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D089725G 25 g
D0897500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D089725G Supplier TCI America Supplier No. D089725G
Only null left

Chemical Identifiers

CAS 101-68-8
Molecular Formula C15H10N2O2
Molecular Weight (g/mol) 250.257
MDL Number MFCD00036131
InChI Key UPMLOUAZCHDJJD-UHFFFAOYSA-N
Synonym 4,4'-diphenylmethane diisocyanate, diphenylmethane diisocyanate, isonate, 4,4'-diisocyanatodiphenylmethane, p,p'-diphenylmethane diisocyanate, bis 4-isocyanatophenyl methane, 4,4'-methylenebis phenyl isocyanate, methylbisphenyl isocyanate, 1,1'-methylenebis 4-isocyanatobenzene, methylene diphenyl diisocyanate
PubChem CID 7570
ChEBI CHEBI:53218
IUPAC Name 1-isocyanato-4-[(4-isocyanatophenyl)methyl]benzene
SMILES C1=CC(=CC=C1CC2=CC=C(C=C2)N=C=O)N=C=O

Specifications

Melting Point 41°C
Color White
Boiling Point 184°C
Quantity 25 g
UN Number 2206
Formula Weight 250.26
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Methylenediphenyl 4,4′-Diisocyanate
EINECSNumber (4)-0118
RTECSNumber NQ9350000
TSCA Yes
Recommended Storage Refrigerator