missing translation for 'onlineSavingsMsg'
Learn More

Methylenediformamide, 97%

Catalog No. AAB2411606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2411606 5 g
AAB2411603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2411606 Supplier Thermo Scientific Chemicals Supplier No. B2411606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6921-98-8 | C3H6N2O2 | 102.093 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6921-98-8
Molecular Formula C3H6N2O2
Molecular Weight (g/mol) 102.093
MDL Number MFCD00021038
InChI Key QPJQPYQZFKFTHG-UHFFFAOYSA-N
Synonym methylenediformamide, n,n'-methylenebisformamide, n,n'-methylenediformamide, methylenebisformamide, formamide, n,n'-methylenebis, n-formamidomethyl formamide, n,n'-methylenebis formamide, methylene-bis-formamide, n,n-methylenebisformamide, acmc-1b7ve
PubChem CID 81339
IUPAC Name N-(formamidomethyl)formamide
SMILES C(NC=O)NC=O

Specifications

Melting Point 141°C to 143°C
Quantity 5 g
Beilstein 1811705
Formula Weight 102.09
Percent Purity 97%
Chemical Name or Material Methylenediformamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 230-041-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only