CAS RN 6921-98-8
IUPAC Name:
N-(formamidomethyl)formamide
Synonyms:
N-(formamidomethyl)formamide
Molecular Weight (g/mol):
102.09
Molecular Formula:
C3H6N2O2
InChi Key:
QPJQPYQZFKFTHG-UHFFFAOYSA-N
SMILES:
O=CNCNC=O
1
–
1
of
1
results
1
Promotions Available
Methylenediformamide, 97%
Methylenediformamide, 97%
CAS: 6921-98-8 Molecular Formula: C3H6N2O2 Molecular Weight (g/mol): 102.093 MDL Number: MFCD00021038 InChI Key: QPJQPYQZFKFTHG-UHFFFAOYSA-N Synonym: methylenediformamide,n,n'-methylenebisformamide,n,n'-methylenediformamide,methylenebisformamide,formamide, n,n'-methylenebis,n-formamidomethyl formamide,n,n'-methylenebis formamide,methylene-bis-formamide,n,n-methylenebisformamide,acmc-1b7ve PubChem CID: 81339 IUPAC Name: N-(formamidomethyl)formamide SMILES: C(NC=O)NC=O