missing translation for 'onlineSavingsMsg'
Learn More

Methylenebis(thioglycolic Acid) 96.0+%, TCI America™

Catalog No. M02185G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M02185G 5 g
M021825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M02185G Supplier TCI America Supplier No. M02185G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2068-24-8
Molecular Formula C5H8O4S2
Molecular Weight (g/mol) 196.235
MDL Number MFCD00135949
InChI Key CZEVAMMETWVBOD-UHFFFAOYSA-N
Synonym methylenedithiodiacetic acid, methylenebisthioglycolic acid, acetic acid, 2,2'-methylenebis thio bis, acetic acid, methylenedithio di, 2,2'-methylenebis thio diacetic acid, methylenebis-thioglycollic acid, 2,2'-methylenebis sulfanediyl diacetic acid, 2-carboxymethyl thio methyl thio acetic acid, carboxymethyl sulfanyl methyl sulfanyl acetic acid
PubChem CID 74949
IUPAC Name 2-(carboxymethylsulfanylmethylsulfanyl)acetic acid
SMILES C(C(=O)O)SCSCC(=O)O

Specifications

Melting Point 129°C
Quantity 5 g
Formula Weight 196.24
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Methylenebis(thioglycolic Acid)