missing translation for 'onlineSavingsMsg'
Learn More

Methylenebis[dimethyl(2-pyridyl)silane] 95.0+%, TCI America™

Catalog No. M1460100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1460100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1460100MG Supplier TCI America Supplier No. M1460100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 243468-48-6
Molecular Formula C15H22N2Si2
Molecular Weight (g/mol) 286.525
MDL Number MFCD06797120
InChI Key WZXVWYFOAXHQGL-UHFFFAOYSA-N
Synonym Bis[dimethyl(2-pyridyl)silyl]methane, 2,2′C-[Methylenebis(dimethylsilylene)]bispyridine
PubChem CID 11196941
IUPAC Name [dimethyl(pyridin-2-yl)silyl]methyl-dimethyl-pyridin-2-ylsilane
SMILES C[Si](C)(C[Si](C)(C)C1=CC=CC=N1)C2=CC=CC=N2

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 286.53
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Methylenebis[dimethyl(2-pyridyl)silane]