accessibility menu, dialog, popup

UserName

CH3SiH3CSiCH3H3CNN

CAS RN 243468-48-6

IUPAC Name: 2-({[dimethyl(pyridin-2-yl)silyl]methyl}dimethylsilyl)pyridine
Synonyms: 2-({[dimethyl(pyridin-2-yl)silyl]methyl}dimethylsilyl)pyridine
Molecular Weight (g/mol): 286.53
Molecular Formula: C15H22N2Si2
InChi Key: WZXVWYFOAXHQGL-UHFFFAOYSA-N
SMILES: C[Si](C)(C[Si](C)(C)C1=CC=CC=N1)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results