missing translation for 'onlineSavingsMsg'
Learn More

Methylene Dithiocyanate 99.0+%, TCI America™

Catalog No. M102025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M102025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M102025G Supplier TCI America Supplier No. M102025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6317-18-6
Molecular Formula C3H2N2S2
Molecular Weight (g/mol) 130.18
MDL Number MFCD00001833
InChI Key JWZXKXIUSSIAMR-UHFFFAOYSA-N
Synonym methylene dithiocyanate, methylene bis thiocyanate, methylenedithiocyanate, proxel mb, methylenedirhodanide, thiocyanic acid, methylene ester, nalfloc n 206, methylenedirhodanid, methylene thiocyanate, dithiocyanatomethane
PubChem CID 22772
IUPAC Name {[(cyanosulfanyl)methyl]sulfanyl}carbonitrile
SMILES N#CSCSC#N

Specifications

Melting Point 105°C
Quantity 25 g
UN Number 2928
Formula Weight 130.18
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methylene Dithiocyanate
EINECSNumber (2)-1693
RTECSNumber XL1560000
TSCA Yes