missing translation for 'onlineSavingsMsg'
Learn More

Methylene Chloride (Stabilized/Spectranalyzed™), Fisher Chemical™

Catalog No. p-216236
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
D351 1 L Glass Bottle
D354 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D351 Supplier Thermo Fisher Scientific Supplier No. D35 1
Only null left

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97°C
Absorbance Pass Test
Color Colorless
Density 1.33g/cm³
Quantity 1 L
Identification Pass Test
Infrared Spectrum Conforms to reference
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.002% max.
Assay Percent Range ≥99.5 %
Titratable Acid 0.0003mEq/g max.
Vapor Pressure 20mmHg at 3502°C
UN Number 1593
Water 0.02% max.
Boiling Point 39°C
Formula Weight 84.93
Percent Purity ≥99.5%
Grade Also meets ACS specifications
Chemical Name or Material Methylene Chloride, Stabilized
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. May be harmful if inhaled. May cause irritation of respiratory tract. Inhalation may cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:0
DOTInformation DOT Class 6.1, : Poison