missing translation for 'onlineSavingsMsg'
Learn More

Methylene Chloride (Not Stabilized/HPLC), Fisher Chemical™

Catalog No. D150SK4
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass
Safe-Cote™ Amber Glass 
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
D150SK4 4 L Safe-Cote™ Amber Glass 
D1501 1 L Amber Glass
D150SK1 1 L Safe-Cote™ Amber Glass 
D1504 4 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. D150SK4 Supplier Thermo Fisher Scientific Supplier No. FLD150SK4
Get 1 Extra Chemical When You Buy 2
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol

Methylene Chloride (Not Stabilized/HPLC)

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97°C
Absorbance 0.01 AU max. at 254nm, 0.20 AU max. at 240nm, 0.70 AU max. at 235nm, 1.0 AU max. at 232nm
Color Colorless
Acidity 0.0005 % max.
Density 1.33g/cm³
Quantity 4 L
Identification Pass test
Packaging Safe-Cote™ Amber Glass 
Physical Form Liquid
Refractive Index 1.4200 to 1.4230
Residue after Evaporation 4ppm max.
Assay Percent Range 99.9% min by GC
Vapor Pressure 20mmHg at 3502°C
Water 0.02% Max.
Boiling Point 39°C
Formula Weight 84.93
Percent Purity ≥99.9%
Grade Suitable for HPLC applications
Chemical Name or Material Methylene Chloride, Not Stabilized
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. May be harmful if inhaled. May cause irritation of respiratory tract. Inhalation may cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:0
DOTInformation DOT Class 6.1, : Poison