missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylene Chloride (Not Stabilized/HPLC), Fisher Chemical™
Description
Methylene Chloride (Not Stabilized/HPLC)
Specifications
Specifications
| Melting Point | -97°C |
| Absorbance | 0.01 AU max. at 254nm, 0.20 AU max. at 240nm, 0.70 AU max. at 235nm, 1.0 AU max. at 232nm |
| Color | Colorless |
| Acidity | 0.0005 % max. |
| Density | 1.33g/cm³ |
| Quantity | 4 L |
| Identification | Pass test |
| Packaging | Amber Glass |
| Physical Form | Liquid |
| Refractive Index | 1.4200 to 1.4230 |
| Show More |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Possible cancer hazard. May cause cancer based on animal data. May be harmful if inhaled. May cause irritation of respiratory tract. Inhalation may cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. NFPA Health:2 Flammability:1 Instability:0 |
| DOTInformation | DOT Class 6.1, : Poison |