missing translation for 'onlineSavingsMsg'
Learn More

Methylene Chloride/Methanol, 98/2 (w/w) (L-038), Reagents

Catalog No. 01334725
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334725 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334725 Supplier Labchem Inc Supplier No. CM1250004C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Methylene Chloride/Methanol, 98/2 (w/w) (L-038), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 75-09-2 , 67-56-1
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym Dichloromethane/Methyl Alcohol
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Color Colorless
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 1.325
Grade Reagent
Chemical Name or Material Methylene Chloride/Methanol, 98/2 (w/w) (L-038)