missing translation for 'onlineSavingsMsg'
Learn More

Methylene Chloride, Hydrocarbon Stabilized, Exceeds A.C.S. Specifications, HPLC Grade, 99.9%, Spectrum™ Chemical

Catalog No. p-7157586
Change view
Click to view available options
Quantity:
4 L
4 x 4 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604510 4 L Amber Glass Bottle
18604512 4 x 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18604510 Supplier Spectrum Chemical Mfg Cor Supplier No. HP7324LT
Only null left

HP732, 75-09-2, CH2Cl2

Spectrum™ Chemical Methylene Chloride, Hydrocarbon Stabilized, Exceeds A.C.S. Specifications, HPLC Grade , otherwise referred to as Dichloromethane. This organic compound has a light sweet aroma and is widely used as a solvent. Although this colorless, volatile liquid is non-miscible with water, it is miscible with a rang of other organic solvents. Thanks to it′s low boiling point, it is able to function in a heat engine. It also welds certain plastics together and is used by hobbyist for joining components together. As it exceeds ACS grade Reagent standards, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 4 L
UN Number UN1593
Residue after Evaporation 4 ppm
Percent Purity 99.9%
Grade High Purity Solvent