missing translation for 'onlineSavingsMsg'
Learn More

Methylene Chloride (HPLC), Fisher Chemical™

Catalog No. p-216218
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Safe-Cote™ Amber Glass 
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
D1431 1 L Glass Bottle
D1434 4 L Glass Bottle
D143SK4 4 L Safe-Cote™ Amber Glass 
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1431 Supplier Thermo Fisher Scientific Supplier No. D1431
Only null left

HPLC mobile phase for analytical and preparative separations.

Contains approx. 15-200 ppm preservative

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97°C
Absorbance 0.01 AU max., 0.01 AU max. at 260nm, 0.014 AU max. at 254nm, 0.10 max. at 250nm, 0.2 AU max. at 240nm, 1 AU max. at 235nm, Pass Test
Color Colorless
Boiling Point 39°C
Quantity 1 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 2ppm max.
Assay Percent Range 99.9% min by GC
Titratable Acid 0.0003mEq/g max.
Vapor Pressure 20mmHg at 3502°C
UN Number 1593
Water 0.02% max.
Formula Weight 84.93
Percent Purity ≥99.9%
Grade Mobile phase for HPLC applications
Chemical Name or Material Methylene Chloride, Stabilized
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. May be harmful if inhaled. May cause irritation of respiratory tract. Inhalation may cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:0
DOTInformation DOT Class 6.1, : Poison