missing translation for 'onlineSavingsMsg'
Learn More

Methylene Chloride, FCC, 99%, Spectrum™ Chemical

Catalog No. 18604503
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Steel Can
Steel Drum
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604503 6 x 1 L -
18604500 100 mL -
18604506 500 mL -
18604501 1 L -
18604507 6 x 500 mL -
18604502 4 L -
18604504 4 x 4 L -
18604505 20 L Steel Can
18604508 200 L Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 18604503 Supplier Spectrum Chemical Mfg Cor Supplier No. M11721LTCS6
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

M1172, 75-09-2, CH2Cl2

Spectrum™ Chemical Methylene Chloride, FCC is used to decaffeinate coffee and tea as well as to prepare extracts of hops and other flavorings. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Acidity 10 mg/kg
CAS Max % 100%
Quantity 6 x 1 L
Specific Gravity 1.318-1.323
Water 0.02%
UN Number UN1593
Boiling Point 39.8°C
Percent Purity 99%
Chemical Name or Material Methylene Chloride