missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue Reagent, For Surfactants, Certified, LabChem™

$97.89 - $142.71

Specifications

Absorbance Passes Test
Chemical Name or Material Methylene Blue Reagent
Name Note For Surfactants
CAS Max % 94.967,4.35,0.68,0.003
CAS 7558-80-7 , 7664-93-9 , 61-73-4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC169701
View Documents
LabChem
LC169701
500 mL
Each for $97.89
Only null left
 
LC169702
View Documents
LabChem
LC169702
1 L
Each for $142.71
Only null left
 
Description

Description

Specifications

Specifications

Passes Test
For Surfactants
7558-80-7 , 7664-93-9 , 61-73-4
Soluble in water
319.85
Poly Bottle
Sulfur compounds; Phosphorus oxides
Blue
Methylene Blue Reagent
94.967,4.35,0.68,0.003
C16H18N3SCl
Passes Test
Certified
1g/mL
1g/mL
Liquid
Videos
SDS
Documents

Documents