missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methylene Blue, pure, certified, residual water

Catalog No. AC414245000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC414245000 500 g Glass bottle
AC414240250 25 g Glass bottle
AC414241000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC414245000 Supplier Thermo Scientific Chemicals Supplier No. 414245000

Methylene Blue, C16H24ClN3O3S, CAS Number-7220-79-3, Basic Blue 9, 25g, 1.21 to 1.7, Vapor Pressure: Negligible, 27, V,19, 259, CHEBI:6872, Green, 13 (Air - 1.0), 82% min., 200-515-2, 319.85, Pure, Warning, GHS H Statement: Harmful if swallowed. | CAS: 7220-79-3 | C16H24ClN3O3S | 373.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7220-79-3
Molecular Formula C16H24ClN3O3S
Molecular Weight (g/mol) 373.90
MDL Number MFCD00150008
InChI Key XQAXGZLFSSPBMK-UHFFFAOYSA-M
Synonym Basic Blue 9
PubChem CID 6099
ChEBI CHEBI:6872
IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride
SMILES O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C

Specifications

Melting Point 190.0°C
Absorption Ratio 1.21 to 1.7
Dye Content 82% min.
Beilstein 27, V,19, 259
Merck Index 15, 6132
Solubility Information Solubility in water: 40g/L (20°C). Other solubilities: in methanol: 430g/L
Infrared Spectrum Authentic
Lambda Max 663 to 667nm
Formula Weight 319.85
Grade Pure
Packaging Glass bottle
Vapor Pressure Vapor Pressure: Negligible
Density 13 (Air = 1.0)
Color Green
Quantity 500 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Methylene Blue
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-515-2
RTECSNumber SO5600000
TSCA TSCA

RUO – Research Use Only