missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Methylene Blue hydrate, pure
CAS: 122965-43-9 | C16H18ClN3S | 319.85 g/mol
$154.60 - $2,198.09
Chemical Identifiers
| CAS | 122965-43-9 |
|---|---|
| Molecular Formula | C16H18ClN3S |
| Molecular Weight (g/mol) | 319.85 |
| MDL Number | MFCD00150006 |
| InChI Key | CXKWCBBOMKCUKX-UHFFFAOYSA-M |
| Synonym | Basic Blue 9 hydrate, C.I. 52015 hydrate |
| PubChem CID | 16211647 |
| SMILES | [Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC414251000
|
Thermo Scientific Chemicals
414251000 |
100 g | Glass bottle |
N/A
|
|
||||
|
AC414255000
|
Thermo Scientific Chemicals
414255000 |
500 g | Glass bottle |
N/A
|
|
||||
|
AC414250025
|
Thermo Scientific Chemicals
414250025 |
2.5 kg | Plastic Drum |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
| 122965-43-9 | |
| 319.85 | |
| CXKWCBBOMKCUKX-UHFFFAOYSA-M | |
| 16211647 |
| C16H18ClN3S | |
| MFCD00150006 | |
| Basic Blue 9 hydrate, C.I. 52015 hydrate | |
| [Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C |
Specifications
| 100.0°C to 110.0°C | |
| C16H18ClN3S | |
| Basic Blue 9 hydrate, C.I. 52015 hydrate | |
| CXKWCBBOMKCUKX-UHFFFAOYSA-M | |
| [Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C | |
| 319.85 | |
| 22% max. | |
| ≥70% (TiCl3-titration) | |
| Glass bottle | |
| 100 g | |
| Methylene Blue hydrate |
| 122965-43-9 | |
| MFCD00150006 | |
| Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol | |
| Authentic | |
| [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;hydrate | |
| 16211647 | |
| 319.85 | |
| Pure | |
| Green | |
| Fine Crystalline Powder |
Safety and Handling
Warning
RUO – Research Use Only