missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Methylene Blue hydrate, pure
CAS: 122965-43-9 | C16H18ClN3S | 319.85 g/mol
Supplier: Thermo Scientific Chemicals 414250025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorSpecifications
| Methylene Blue hydrate | |
| 122965-43-9 | |
| MFCD00150006 | |
| Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol | |
| Authentic | |
| 7-(dimethylamino)-N,N-dimethyl-3H-phenothiazin-3-iminium chloride | |
| 16211647 | |
| 319.85 | |
| Pure | |
| Green | |
| Fine Crystalline Powder |
| 100.0°C to 110.0°C | |
| C16H18ClN3S | |
| Basic Blue 9 hydrate, C.I. 52015 hydrate | |
| CXKWCBBOMKCUKX-UHFFFAOYSA-M | |
| [Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C | |
| 319.85 | |
| 22% max. | |
| ≥70% (TiCl3-titration) | |
| Plastic Drum | |
| 2.5 kg |
Chemical Identifiers
| 122965-43-9 | |
| 319.85 | |
| CXKWCBBOMKCUKX-UHFFFAOYSA-M | |
| 16211647 |
| C16H18ClN3S | |
| MFCD00150006 | |
| Basic Blue 9 hydrate, C.I. 52015 hydrate | |
| [Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C |
Safety and Handling
Warning
RUO – Research Use Only