missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methylene Blue, high purity, biological stain

CAS: 122965-43-9 | C16H18ClN3S | 319.85 g/mol

$73.42 - $591.38

Chemical Identifiers

CAS 122965-43-9
Molecular Formula C16H18ClN3S
Molecular Weight (g/mol) 319.85
MDL Number MFCD00150006
InChI Key CXKWCBBOMKCUKX-UHFFFAOYSA-M
Synonym Basic Blue 9; C.I. 52015
PubChem CID 16211647
IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;hydrate
SMILES [Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1817414
View Documents
Thermo Scientific Chemicals
A1817414
25 g
Each for $73.42
Only null left
 
AAA1817422
View Documents
Thermo Scientific Chemicals
A1817422
100 g
Each for $157.82
Only null left
 
AAA1817436
View Documents
Thermo Scientific Chemicals
A1817436
500 g
Each for $591.38
Only null left
 
Description

Description

Methylene blue is used as a dye in different staining procedures viz. Wright's stain and Jenner's stain. It also serves as an indicator and medicine. Further, it is used to examine RNA and DNA under the microscope or in a gel. It is widely used as a redox indicator in analytical chemistry. It acts as a photosensitizer utilized to generate singlet oxygen in order to make organic peroxides by a Diels-Alder reaction. It is also used in the sulfide analysis. It plays an important role in aquaculture to treat fungal infections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylene blue is used as a dye in different staining procedures viz. Wright′s stain and Jenner′s stain. It also serves as an indicator and medicine. Further, it is used to examine RNA and DNA under the microscope or in a gel. It is widely used as a redox indicator in analytical chemistry. It acts as a photosensitizer utilized to generate singlet oxygen in order to make organic peroxides by a Diels-Alder reaction. It is also used in the sulfide analysis. It plays an important role in aquaculture to treat fungal infections.

Solubility
Soluble in water, alcohol, acetic acid, glycerol and chloroform. Insoluble in ether.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

122965-43-9
319.85
CXKWCBBOMKCUKX-UHFFFAOYSA-M
16211647
[Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C
C16H18ClN3S
MFCD00150006
Basic Blue 9; C.I. 52015
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;hydrate

Specifications

∼180°C (decomposition)
C16H18ClN3S
3599847
Basic Blue 9; C.I. 52015
CXKWCBBOMKCUKX-UHFFFAOYSA-M
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;hydrate
16211647
25 g
High purity
122965-43-9
MFCD00150006
14,6060
Soluble in water,alcohol,acetic acid,glycerol and chloroform. Insoluble in ether.
[Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C
319.85
319.86 (Anhydrous)
Methylene Blue, biological stain
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 200-515-2

RTECSNumber : SO5600000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only