missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methylene Blue, high purity, biological stain

CAS: 122965-43-9 | C16H18ClN3S | 319.85 g/mol

Supplier:  Thermo Scientific Chemicals A1817422

 View more versions of this product

Catalog No. AAA1817422


Only null left
Explore available promotions

Description

Description

Methylene blue is used as a dye in different staining procedures viz. Wright's stain and Jenner's stain. It also serves as an indicator and medicine. Further, it is used to examine RNA and DNA under the microscope or in a gel. It is widely used as a redox indicator in analytical chemistry. It acts as a photosensitizer utilized to generate singlet oxygen in order to make organic peroxides by a Diels-Alder reaction. It is also used in the sulfide analysis. It plays an important role in aquaculture to treat fungal infections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylene blue is used as a dye in different staining procedures viz. Wright′s stain and Jenner′s stain. It also serves as an indicator and medicine. Further, it is used to examine RNA and DNA under the microscope or in a gel. It is widely used as a redox indicator in analytical chemistry. It acts as a photosensitizer utilized to generate singlet oxygen in order to make organic peroxides by a Diels-Alder reaction. It is also used in the sulfide analysis. It plays an important role in aquaculture to treat fungal infections.

Solubility
Soluble in water, alcohol, acetic acid, glycerol and chloroform. Insoluble in ether.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Methylene Blue
biological stain
High purity
MFCD00150006
14,6060
Soluble in water,alcohol,acetic acid,glycerol and chloroform. Insoluble in ether.
[Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C
319.85
319.86 (Anhydrous)
∼180°C (decomposition)
122965-43-9
C16H18ClN3S
3599847
Basic Blue 9; C.I. 52015
CXKWCBBOMKCUKX-UHFFFAOYSA-M
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;hydrate
16211647
100 g

Chemical Identifiers

122965-43-9
319.85
CXKWCBBOMKCUKX-UHFFFAOYSA-M
16211647
[Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C
C16H18ClN3S
MFCD00150006
Basic Blue 9; C.I. 52015
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;hydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 200-515-2

RTECSNumber : SO5600000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only