missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue Chloride, Spectrum™ Chemical

Catalog No. 18611770
Change view
Click to view available options
Quantity:
25 g
25 kg
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611770 25 g Amber Glass Bottle
18611771 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18611770 Supplier Spectrum Chemical Mfg Cor Supplier No. ME14525GM

Chemical Identifiers

CAS 7220-79-3

Specifications

CAS Max % 1
Grade Reagent
Packaging Amber Glass Bottle
Color Blue/Green
Quantity 25 g
Chemical Name or Material Methylene Blue Chloride

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.