missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue (Certified Biological Stain), Fisher Chemical™

Catalog No. M291100
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M291100 100 g Glass Bottle
M29125 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M291100 Supplier Thermo Fisher Scientific Supplier No. M291 100
Edge

For use in histology, bacteriology (staining), compounding of blood stains

Chemical Identifiers

CAS 7220-79-3
Molecular Formula C16H24ClN3O3S
Molecular Weight (g/mol) 373.90
MDL Number MFCD00150008
InChI Key XQAXGZLFSSPBMK-UHFFFAOYSA-M
Synonym Basic Blue 9
PubChem CID 104827
IUPAC Name 3,7-bis(dimethylamino)-5λ⁴-phenothiazin-5-ylium trihydrate chloride
SMILES O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C

Specifications

Melting Point 190°C
Bio. Stain Comm. Certification Pass Test
Dye Content Report %
Formula Weight 373.9
Grade Certified Biological Stain
Packaging Glass Bottle
Vapor Pressure Vapor Pressure: Negligible
Decomposition Information 190°C
Density 1.230g/cm³
Color Index No. 52015
Color Green
pH 3 to 4.5
Quantity 100 g
Physical Form Solid
Chemical Name or Material Methylene Blue
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Harmful if swallowed. Causes eye, skin, and respiratory tract irritation. May cause methemoglobinemia. Ensure adequate ventilation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:2
Flammability:1
Instability:0