missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylene Blue, C.I. 52015, Spectrum™ Chemical
$329.27 - $7225.13
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18611782
|
Spectrum Chemical Mfg Cor
ME14025GM |
25 g | Amber Glass Bottle |
Each for $329.27
|
|
||||
|
18611767
|
Spectrum Chemical Mfg Cor
ME140100GM |
100 g | Amber Glass Bottle |
Each for $684.40
|
|
||||
|
18611781
|
Spectrum Chemical Mfg Cor
ME1401KGBL |
1 kg | Amber Glass Bottle |
Each for $4,007.90
|
|
||||
|
18611768
|
Spectrum Chemical Mfg Cor
ME14025KGBL |
2.5 kg | Amber Glass Bottle |
Each for $7,225.13
|
|
||||
Chemical Identifiers
| 7220-79-3 |
Specifications
| 7220-79-3 | |
| Reagent | |
| Blue/Green | |
| Methylene Blue, C.I. 52015 |
| 1 | |
| Amber Glass Bottle | |
| 25 g |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.