missing translation for 'onlineSavingsMsg'
Learn More

Methyle3-hydroxybenzo[b]thiophene-2-carboxylate, 97%, Thermo Scientific™

Catalog No. BTB04875DA
Change view
Click to view available options
Quantity:
1g
10g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB04875DA 1g Amber Glass Bottle
BTB04875EA 10g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB04875DA Supplier Thermo Scientific Chemicals Supplier No. BTB04875DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13134-76-4
Molecular Formula C10H8O3S
Molecular Weight (g/mol) 208.23
MDL Number MFCD00067732
InChI Key JVUOTPTULVZUTL-KTKRTIGZSA-N
Synonym methyl 3-hydroxybenzo b thiophene-2-carboxylate, methyl 3-hydroxybenzo b thiophene-3-carboxylate, 2-hydroxy methoxy methylidene-1-benzothiophen-3-one, 2z-2-hydroxy methoxy methylidene-1-benzothiophen-3-one, benzo b thiophene-2-carboxylicacid, 3-hydroxy-, methyl ester, benzo b thiophene-2-carboxylic acid, 3-hydroxy-, methyl ester, maybridge4_000295, methyl 3-hydroxy-benzothiophene2-carboxylate
PubChem CID 83165
IUPAC Name methyl 3-hydroxy-1-benzothiophene-2-carboxylate
SMILES CO\C(O)=C1/SC2=CC=CC=C2C1=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 208.24
Percent Purity 97%
Chemical Name or Material Methyl 3-hydroxybenzo[b]thiophene-2-carboxylate
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.