CAS RN 13134-76-4
IUPAC Name:
(2Z)-2-[hydroxy(methoxy)methylidene]-2,3-dihydro-1-benzothiophen-3-one
Synonyms:
(2Z)-2-[hydroxy(methoxy)methylidene]-1-benzothiophen-3-one
Molecular Weight (g/mol):
208.23
Molecular Formula:
C10H8O3S
InChi Key:
JVUOTPTULVZUTL-KTKRTIGZSA-N
SMILES:
CO\C(O)=C1/SC2=CC=CC=C2C1=O
1
–
1
of
1
results
1
Methyle3-hydroxybenzo[b]thiophene-2-carboxylate, 97%, Thermo Scientific™
CAS: 13134-76-4 Molecular Formula: C10H8O3S Molecular Weight (g/mol): 208.23 MDL Number: MFCD00067732 InChI Key: JVUOTPTULVZUTL-KTKRTIGZSA-N Synonym: methyl 3-hydroxybenzo b thiophene-2-carboxylate,methyl 3-hydroxybenzo b thiophene-3-carboxylate,2-hydroxy methoxy methylidene-1-benzothiophen-3-one,2z-2-hydroxy methoxy methylidene-1-benzothiophen-3-one,benzo b thiophene-2-carboxylicacid, 3-hydroxy-, methyl ester,benzo b thiophene-2-carboxylic acid, 3-hydroxy-, methyl ester,maybridge4_000295,methyl 3-hydroxy-benzothiophene2-carboxylate PubChem CID: 83165 IUPAC Name: (2Z)-2-[hydroxy(methoxy)methylidene]-2,3-dihydro-1-benzothiophen-3-one SMILES: CO\C(O)=C1/SC2=CC=CC=C2C1=O