missing translation for 'onlineSavingsMsg'
Learn More

Methyldiphenylsulfonium tetrafluoroborate, 95%

CAS: 10504-60-6 | C13H13BF4S | 288.11 g/mol

$1694.70 - $1694.70

Chemical Identifiers

CAS 10504-60-6
Molecular Formula C13H13BF4S
Molecular Weight (g/mol) 288.11
MDL Number MFCD00051861
InChI Key LHAMVDBAOJCBDW-UHFFFAOYSA-N
Synonym diphenyl methyl sulfonium tetrafluoroborate, methyldiphenylsulphonium tetrafluoroborate 1-, methyldiphenylsulfonium tetrafluoroborate, methyldiphenylsulfanium tetrafluoroborate, acmc-1bx8v, diphenyl methyl sulfoniumtetrafluoroborate, methyl diphenyl sulfanium tetrafluoroborate, diphenyl methyl sulphonium tetrafluoroborate, methyl diphenyl sulfonium tetrafluoroborate
PubChem CID 2758717
IUPAC Name methyl(diphenyl)sulfanium;tetrafluoroborate
SMILES F[B-](F)(F)F.C[S+](C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0051214
View Documents
Thermo Scientific Chemicals
L0051214
25 g
Each for $1,694.70
Only null left
 
Description

Description

Methyldiphenylsulfonium tetrafluoroborate is a Methylene transfer agent. It is also used in alkyl diphenylsulfonium salts in place of trialkyl avoids sulfur-containing by-products arising from the Sommelet-Hauser rearrangement.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyldiphenylsulfonium tetrafluoroborate is a Methylene transfer agent. It is also used in alkyl diphenylsulfonium salts in place of trialkyl avoids sulfur-containing by-products arising from the Sommelet-Hauser rearrangement.

Solubility
Soluble in water, alcohol.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications

Chemical Identifiers

10504-60-6
288.11
LHAMVDBAOJCBDW-UHFFFAOYSA-N
2758717
F[B-](F)(F)F.C[S+](C1=CC=CC=C1)C1=CC=CC=C1
C13H13BF4S
MFCD00051861
diphenyl methyl sulfonium tetrafluoroborate, methyldiphenylsulphonium tetrafluoroborate 1-, methyldiphenylsulfonium tetrafluoroborate, methyldiphenylsulfanium tetrafluoroborate, acmc-1bx8v, diphenyl methyl sulfoniumtetrafluoroborate, methyl diphenyl sulfanium tetrafluoroborate, diphenyl methyl sulphonium tetrafluoroborate, methyl diphenyl sulfonium tetrafluoroborate
methyl(diphenyl)sulfanium;tetrafluoroborate

Specifications

10504-60-6
C13H13BF4S
MFCD00051861
UN1759
diphenyl methyl sulfonium tetrafluoroborate, methyldiphenylsulphonium tetrafluoroborate 1-, methyldiphenylsulfonium tetrafluoroborate, methyldiphenylsulfanium tetrafluoroborate, acmc-1bx8v, diphenyl methyl sulfoniumtetrafluoroborate, methyl diphenyl sulfanium tetrafluoroborate, diphenyl methyl sulphonium tetrafluoroborate, methyl diphenyl sulfonium tetrafluoroborate
LHAMVDBAOJCBDW-UHFFFAOYSA-N
methyl(diphenyl)sulfanium;tetrafluoroborate
2758717
95%
63°C to 67°C
(C6H5)2SCH3BF4
25 g
4732019
Soluble in water,alcohol.
F[B-](F)(F)F.C[S+](C1=CC=CC=C1)C1=CC=CC=C1
288.11
288.11
Methyldiphenylsulfonium tetrafluoroborate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

EINECSNumber : 234-032-3

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only