accessibility menu, dialog, popup

UserName

H3CSFBFFF

CAS RN 10504-60-6

IUPAC Name: methyldiphenylsulfanium; tetrafluoroboranuide
Synonyms: methyldiphenylsulfanium tetrafluoroborate
Molecular Weight (g/mol): 288.11
Molecular Formula: C13H13BF4S
InChi Key: LHAMVDBAOJCBDW-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.C[S+](C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)