missing translation for 'onlineSavingsMsg'
Learn More

Methyldiphenylphosphine 97.0+%, TCI America™

Catalog No. p-7132660
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13181G 1 g
M13185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13181G Supplier TCI America Supplier No. M13181G
Only null left

Chemical Identifiers

CAS 1486-28-8
Molecular Formula C13H13P
Molecular Weight (g/mol) 200.22
MDL Number MFCD00008508
InChI Key UJNZOIKQAUQOCN-UHFFFAOYSA-N
Synonym methyldiphenylphosphine, diphenylmethylphosphine, methyl diphenyl phosphine, phosphine, methyldiphenyl, methyldiphenylphsphine, diphenylphosphinomethane, methyldiphenylphoshine, pubchem6421, methyl diphenylphosphine, methyldi-phenylphosphine
PubChem CID 73879
IUPAC Name methyldiphenylphosphane
SMILES CP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 186°C
Quantity 1 g
Formula Weight 200.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyldiphenylphosphine