accessibility menu, dialog, popup

UserName

CAS RN 1486-28-8

IUPAC Name: methyldiphenylphosphane
Synonyms: methyldiphenylphosphane
Molecular Weight (g/mol): 200.22
Molecular Formula: C13H13P
InChi Key: UJNZOIKQAUQOCN-UHFFFAOYSA-N
SMILES: CP(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)