missing translation for 'onlineSavingsMsg'
Learn More

Methyldiphenylphosphine, 99%

Catalog No. AC408970050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC408970050 5 g Glass bottle
AC408970250 25 g Glass bottle
AC408971000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC408970050 Supplier Thermo Scientific Chemicals Supplier No. 408970050

CAS: 1486-28-8 | C13H13P | 200.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1486-28-8
Molecular Formula C13H13P
Molecular Weight (g/mol) 200.22
MDL Number MFCD00008508
InChI Key UJNZOIKQAUQOCN-UHFFFAOYSA-N
Synonym methyldiphenylphosphine, diphenylmethylphosphine, methyl diphenyl phosphine, phosphine, methyldiphenyl, methyldiphenylphsphine, diphenylphosphinomethane, methyldiphenylphoshine, pubchem6421, methyl diphenylphosphine, methyldi-phenylphosphine
PubChem CID 73879
IUPAC Name methyl(diphenyl)phosphane
SMILES CP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Colorless to Yellow
Density 1.0760g/mL
Boiling Point 118.0°C to 120.0°C (2.2 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6240 to 1.6260
Linear Formula (C6H5)2PCH3
Quantity 5 g
Beilstein 16, II, 371
Specific Gravity 1.076
Solubility Information Solubility in water: insoluble
Formula Weight 200.22
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyldiphenylphosphine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-065-5

RUO – Research Use Only