missing translation for 'onlineSavingsMsg'
Learn More

Methylcyclopentane, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187479
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019124 10 mL
111018218 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019124 Supplier Merck Emd Millipore Supplier No. 6649010ML
Only null left

Methylcyclopentane, a naphthene hydrocarbon, has a tertiary carbon atom and also has greater reactivity towards chemical agents like sulphuric acid, fuming nitric acid, etc.

Featured Industries: Environmental

Specifications

CAS 96-37-7
Ignition Point 624°F
Vapor Pressure 232.8 mmHg (37.7°C)
Density 0.748 g/mL (at 20°C)
Flash Point -29.0°C - closed cup
Physical Form Neat
Linear Formula C5H9 CH3
MDL Number MFCD00001382
Melting Point -142°C
Boiling Point 72°C
Type Analytical Standard
Quantity 10 mL
Formula Weight 84.16
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less