missing translation for 'onlineSavingsMsg'
Learn More

Methylcyclopentadiene dimer, 90%, stabilized

CAS: 26472-00-4 | C12H16 | 160.26 g/mol

Supplier:  Thermo Scientific Chemicals 156850010

 View more versions of this product

Catalog No. AC156850010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methylcyclopentadiene dimer
-51.0°C
0.9400g/mL
26°C
90%
C12H16
MFCD00082297
0.94
160.26
95%
26472-00-4
Colorless to Yellow
200.0°C
Authentic
Glass bottle
1.4975 to 1.4995
1 kg
Solubility in water: insoluble. Other solubilities: soluble in alcohol,benzene and ether
160.26
Liquid

Chemical Identifiers

26472-00-4
C12H16
160.26
MFCD00082297
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Suspected of causing cancer.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 247-724-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only