missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylcyclopentadiene dimer, 90%, stabilized
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -51.0°C |
| Color | Colorless to Yellow |
| Density | 0.9400g/mL |
| Boiling Point | 200.0°C |
| Flash Point | 26°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 90% |
| Packaging | Glass bottle |
| Refractive Index | 1.4975 to 1.4995 |
| Quantity | 250 g |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Flammable liquid and vapour. Suspected of causing cancer. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Obtain special instructions before use. Use personal protective equipment as required. IF exposed or concerned: Get medical advice/attention. |
| EINECSNumber | 247-724-5 |
RUO – Research Use Only