missing translation for 'onlineSavingsMsg'
Learn More

Methylcyclohexane, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020574
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020574 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020574 Supplier Merck Emd Millipore Supplier No. 662941ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Methylcyclohexane [MCH] is a clear organic flammable liquid, mostly used as a solvent. During the development of chemical kinetic mechanisms for other cycloalkanes, MCH is considered as reaction starting point.

Featured Industries: Environmental

Specifications

CAS 108-87-2
Ignition Point 545°F
Vapor Pressure 83.2 mmHg (37.7°C)
Vapor Density 3.4 (vs. air)
Density 0.77 g/mL (at 25°C)
Flash Point -4.0°C - closed cup
Physical Form Neat
Linear Formula C6H11 CH3
MDL Number MFCD00001497
Melting Point -126°C
Boiling Point 101°C
Synonym Hexahydrotoluene
Type Analytical Standard
Quantity 1 mL
Formula Weight 98.19
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less