missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylcellulose, USP, 26-33%, Spectrum™ Chemical
M1314, 9004-67-5
$688.17 - $16760.89
Chemical Identifiers
| CAS | 9004-67-5 |
|---|---|
| Molecular Formula | C20H38O11 |
| Molecular Weight (g/mol) | 454.51 |
| InChI Key | YLGXILFCIXHCMC-UHFFFAOYNA-N |
| IUPAC Name | 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane |
| SMILES | COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18604480
|
Spectrum Chemical Mfg Cor
M1314500GM |
500 g | Poly Pail |
Each for $688.17
|
|
||||
|
18604481
|
Spectrum Chemical Mfg Cor
M131425KG |
2.5 kg | Poly Pail |
Each for $2,509.49
|
|
||||
|
18604482
|
Spectrum Chemical Mfg Cor
M131412KG |
12 kg | Fiber Drum |
Each for $5,861.09
|
|
||||
|
18604483
|
Spectrum Chemical Mfg Cor
M131445KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Methylcellulose, USP is a bulk-forming laxative. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 9004-67-5 | |
| 454.51 | |
| 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane |
| C20H38O11 | |
| YLGXILFCIXHCMC-UHFFFAOYNA-N | |
| COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC |
Specifications
| 9004-67-5 | |
| 1.5% | |
| Poly Pail | |
| 500 g | |
| COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC | |
| 454.51 | |
| USP |
| 100% | |
| 5% | |
| C20H38O11 | |
| YLGXILFCIXHCMC-UHFFFAOYNA-N | |
| 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane | |
| 26 to 33% |