missing translation for 'onlineSavingsMsg'
Learn More

Methylcellulose, 400 cP, USP, 26-33%, Spectrum™ Chemical

Catalog No. 18604477
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604477 12 kg Fiber Drum
18604474 100 g Poly Bottle
18604475 500 g Poly Bottle
18604476 2.5 kg Poly Pail
18604478 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18604477 Supplier Spectrum Chemical Mfg Cor Supplier No. ME13612KG
Only null left

ME136, 9004-67-5

Spectrum™ Chemical Methylcellulose, 400 cP, USP is a bulk-forming laxative. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9004-67-5
Molecular Formula C20H38O11
Molecular Weight (g/mol) 454.51
InChI Key YLGXILFCIXHCMC-UHFFFAOYNA-N
IUPAC Name 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane
SMILES COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC

Specifications

CAS Max % 100%
Ignition Residue 1.5%
Loss on Drying 5%
Packaging Fiber Drum
Quantity 12 kg
Percent Purity 26 to 33%
Grade USP