missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylcellulose, 400 cP, USP, 26-33%, Spectrum™ Chemical
ME136, 9004-67-5
Supplier: Spectrum Chemical Mfg Cor ME13625KG
Description
Spectrum™ Chemical Methylcellulose, 400 cP, USP is a bulk-forming laxative. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 9004-67-5 | |
| 1.5% | |
| Poly Pail | |
| 2.5 kg | |
| COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC | |
| 454.51 | |
| USP |
| 100% | |
| 5% | |
| C20H38O11 | |
| YLGXILFCIXHCMC-UHFFFAOYNA-N | |
| 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane | |
| 26 to 33% |
Chemical Identifiers
| 9004-67-5 | |
| 454.51 | |
| 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane |
| C20H38O11 | |
| YLGXILFCIXHCMC-UHFFFAOYNA-N | |
| COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC |