missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylcellulose, 4,000 cP, FCC, 27.5-31.5%, Spectrum™ Chemical
M1164, 9004-67-5
Supplier: Spectrum Chemical Mfg Cor M116425KG
Description
Spectrum™ Chemical Methylcellulose, 4,000 cP, FCC is often used as a thickener and emulsifier. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, personal care and nutritional supplement applications. It is added to hair shampoos, tooth pastes and liquid soaps, to generate their characteristic thick consistency. This is also done for foods in addition to its emulsification properties. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 9004-67-5 | |
| 1.5% | |
| Poly Pail | |
| 2.5 kg | |
| COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC | |
| 454.51 | |
| FCC |
| 100% | |
| 5% | |
| C20H38O11 | |
| YLGXILFCIXHCMC-UHFFFAOYNA-N | |
| 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane | |
| 27.5 to 31.5% |
Chemical Identifiers
| 9004-67-5 | |
| 454.51 | |
| 3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane |
| C20H38O11 | |
| YLGXILFCIXHCMC-UHFFFAOYNA-N | |
| COCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC |