missing translation for 'onlineSavingsMsg'
Learn More

Methylaminoacetonitrile 98.0+%, TCI America™

Catalog No. p-7132014
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M01405ML 5 mL
M014025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M01405ML Supplier TCI America Supplier No. M01405ML
Only null left

Chemical Identifiers

CAS 5616-32-0
Molecular Formula C3H6N2
Molecular Weight (g/mol) 70.095
MDL Number MFCD00058964
InChI Key PVVRRUUMHFWFQV-UHFFFAOYSA-N
PubChem CID 36652
IUPAC Name 2-(methylamino)acetonitrile
SMILES CNCC#N

Specifications

Color Yellow
Boiling Point 65°C
Quantity 5 mL
UN Number 1993
Formula Weight 70.10
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Methylaminoacetonitrile
EINECSNumber (9)-1465
TSCA Yes
Recommended Storage Freezer